Learn More
Hach Company COD Digestion Vials, High Range Plus, 200 to 15,000 mg/L COD, pk/150
Supplier:  Hach Company 2415915
Encompass
For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion method. Hach Method 8000. High Range Plus: 200 to 15,000 mg/L COD. This range is not USEPA Approved. Pack of 150 vials.
Catalog No. 50-199-5957