Learn More
Hach Company COD Digestion Vials, High Range Plus, 200 to 15,000 mg/L COD, pk/25
Catalog No. 501995958 Shop All Hach Company Products
Encompass
For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion method. Hach Method 8000. High Range Plus: 200 to 15,000 mg/L COD. THIS METHOD IS NOT USEPA APPROVED. Pack of 25 vials.
This item is not returnable. View return policy
Catalog No. 50-199-5958 Supplier Hach Company Supplier No. 2415925
For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion method. Hach Method 8000. High Range Plus: 200 to 15,000 mg/L COD. THIS METHOD IS NOT USEPA APPROVED. Pack of 25 vials.