Learn More
Hach Company COD Digestion Vials, Ultra Low Range, 1 to 40 mg/L COD, pk/150
Catalog No. 501995955 Shop All Hach Company Products
Encompass
For Ultra Low Range Chemical Oxygen Demand (COD) determination by the Reactor Digestion Method. Hach Method 8000. Range: 0.7 - 40.0 mg/L COD. Pack of 150 vials. NOTE: This method is not USEPA approved.
Catalog No. 50-199-5955 Supplier Hach Company Supplier No. 2415815
For Ultra Low Range Chemical Oxygen Demand (COD) determination by the Reactor Digestion Method. Hach Method 8000. Range: 0.7 - 40.0 mg/L COD. Pack of 150 vials. NOTE: This method is not USEPA approved.