Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cod Liver Oil, USP, Spectrum™ Chemical
SDP

Catalog No. 18601799 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-799 500 mL Amber Glass Bottle
18-601-800 4 L Amber Glass Bottle
18-601-801 4 x 4 L Amber Glass Bottle
18-601-802 20 L Poly Drum
18-601-803 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-799 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CO112500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CO112, 8001-69-2

Spectrum™ Chemical Cod Liver Oil, USP is an oil from the liver of Cod fish. It is one of the best sources of omega 3 fatty acids (EPA and DHA) and contains relatively high amounts of vitamin A and vitamin D. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-69-2

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.