Learn More
LaMotte™ COD Multi-Range Reagent Systems Vials
SDPSCI_ED
Catalog No. s43593 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
25 ready-to-use vials
Catalog No. Quantity
S43593 25 ready-to-use vials
1 options
Catalog No. S43593 Supplier LaMotte™ Supplier No. 0074SC
Add to Cart
Add to Cart
Lamotte™ Chemical oxygen demand reagent systems are an easy and precise way to measure critical COD levels.
  • Range: 0 to 15,000ppm (High)
  • Mercury-free
Specifications
For Use With (Equipment) Lamotte™ COD PLUS Colorimeter; SMART 2 Colorimeter, SMART Spectro Spectrophotometer
Quantity 25 ready-to-use vials
Product Type COD Reagent System
WARNING: Cancer - www.P65Warnings.ca.gov