Learn More
HANNA INSTRUMENTS INC COD Test Tube Heater Thermoreactor
SDP
Catalog No. NC1862668
Encompass
The HI839800 COD Test Tube Heater is an easy to use thermoreactor constructed of durable materials. The aluminum block incorporates a 25 vial capacity and a well for a reference temperature probe. Its well-marked user interface provides intuitive operation, equipped with two arrows to set the built-in timer and a temperature button with two predefined temperature settings: 150C and 105C. Additional features include status indicator lights, a thermal fuse to prevent overheating, and continuous LCD display.Low and high temperature alertsBuilt-in countdown timerStatus indicator lights
Catalog No. NC1862668 Supplier HANNA INSTRUMENTS INC Supplier No. HI83980001
The HI839800 COD Test Tube Heater is an easy to use thermoreactor constructed of durable materials. The aluminum block incorporates a 25 vial capacity and a well for a reference temperature probe. Its well-marked user interface provides intuitive operation, equipped with two arrows to set the built-in timer and a temperature button with two predefined temperature settings: 150C and 105C. Additional features include status indicator lights, a thermal fuse to prevent overheating, and continuous LCD display.Low and high temperature alertsBuilt-in countdown timerStatus indicator lights