Learn More
Kent Scientific CODA™ Animal Holder
SDP
Catalog No. 13005162 Shop All Kent Scientific Products
Encompass
Change view
Click to view available options
For Use With (Equipment):
Mouse
Rat
Size:
Small
Medium
Large
X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Size
13-005-162 Rat X-Large
13-005-156 Mouse Small
13-005-157 Mouse Medium
13-005-158 Mouse X-Large
13-005-159 Rat Small
13-005-160 Rat Medium
13-005-161 Rat Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-005-162 Supplier Kent Scientific Supplier No. HLDRX
Allows researchers to constantly observe animal behavior during animal research.
  • Adjustable nose cone limits movement
  • Helps animal remain comfortable
  • Decreases injury to animal and researcher
  • Dark black nose cone helps animal feel secure
  • Nose protrudes through cone front, tail exits through rear hatch opening
Specifications
Construction Adjustable black nose cone
For Use With (Application) Observe Animal Behavior During Research
Type Animal Holder
For Use With (Equipment) Rat
Size X-Large