Learn More
Perkin Elmer US LLC Universal Coded Intensitron Lamp Plug Adapter for AAnalyst
Catalog No. NC1928559
Encompass_Preferred
Allows coded lamps (N06612XX series, 12-pin) to be used with AAnalyst instruments (4-pin); however, an AAnalyst spectrometer does not recognize their coding.
Catalog No. NC1928559 Supplier Perkin Elmer US LLC Supplier No. N3050196
Allows coded lamps (N06612XX series, 12-pin) to be used with AAnalyst instruments (4-pin); however, an AAnalyst spectrometer does not recognize their coding.