Learn More
Cerilliant Corporation CODEINE-D6
Catalog No. NC9657215
Encompass
Codeine-D6, 100 ug/mL; 1 mL/ampoule
Catalog No. NC9657215 Supplier Cerilliant Corporation Supplier No. C040
Codeine-D6, 100 ug/mL; 1 mL/ampoule