Learn More
Quanterix Inc CODEX ASSAY REAGENT
SDP
Catalog No. NC1663057
Encompass
  • Facilitates detection of DNA-conjugated antibodies
  • Compatible with formalin-fixed paraffin-embedded (FFPE) tissues
  • Compatible with fresh-frozen (FF) tissues
  • Supports human, mouse, and monkey tissue samples
  • Provides single-cell resolution
  • Maintains spatial context of tissue
  • Enables scalable experiments
  • Allows customizable antibody panels
  • Compatible with PhenoCycler (CODEX) platform
  • Supports tissue microarrays with specimens up to 1 cm²
  • Unit size: 1.25 mL
This item is not returnable. View return policy
Catalog No. NC1663057 Supplier Quanterix Inc Supplier No. 7000002
  • Facilitates detection of DNA-conjugated antibodies
  • Compatible with formalin-fixed paraffin-embedded (FFPE) tissues
  • Compatible with fresh-frozen (FF) tissues
  • Supports human, mouse, and monkey tissue samples
  • Provides single-cell resolution
  • Maintains spatial context of tissue
  • Enables scalable experiments
  • Allows customizable antibody panels
  • Compatible with PhenoCycler (CODEX) platform
  • Supports tissue microarrays with specimens up to 1 cm²
  • Unit size: 1.25 mL