Learn More
Aldevron Codex HiCap RNA Polymerase 10 mL
Catalog No. NC3900951
Encompass_Preferred
Codex HiCap RNA Polymerase is a co-transcriptional capping RNA polymerase engineered to produce synthetic RNA to meet increasing demands for safety and cost-effectiveness. The HiCap enzyme generates fewer undesirable double-stranded RNA (dsRNA) byproducts than wild-type T7 (WT T7) RNA polymerase, reducing unwanted immunogenicity, while efficiently incorporating commercially available cap analogs.
Catalog No. NC3900951 Supplier Aldevron Supplier No. 914510ML
Add to Cart
Add to Cart
Request a Quote
Codex HiCap RNA Polymerase is a co-transcriptional capping RNA polymerase engineered to produce synthetic RNA to meet increasing demands for safety and cost-effectiveness. The HiCap enzyme generates fewer undesirable double-stranded RNA (dsRNA) byproducts than wild-type T7 (WT T7) RNA polymerase, reducing unwanted immunogenicity, while efficiently incorporating commercially available cap analogs.