Learn More
Aldevron Codex HiCapRNA Polymerase, 25uL
Catalog No. 503073867
Encompass_Preferred
25uL dispense of HiCap T7 RNA Polymerase. Codex HiCap RNA Polymerase is a co-transcriptional capping RNA polymerase engineered to produce synthetic RNA to meet increasing demands for safety and cost-effectiveness. The HiCap enzyme generates fewer undesirable double-stranded RNA (dsRNA) byproducts than wild-type T7 (WT T7) RNA polymerase, reducing unwanted immunogenicity, while efficiently incorporating commercially available cap analogs.
Catalog No. 50-307-3867 Supplier Aldevron Supplier No. 914525UL
25uL dispense of HiCap T7 RNA Polymerase. Codex HiCap RNA Polymerase is a co-transcriptional capping RNA polymerase engineered to produce synthetic RNA to meet increasing demands for safety and cost-effectiveness. The HiCap enzyme generates fewer undesirable double-stranded RNA (dsRNA) byproducts than wild-type T7 (WT T7) RNA polymerase, reducing unwanted immunogenicity, while efficiently incorporating commercially available cap analogs.