Learn More
Aldevron Codex HiCapRNA Polymerase, 1mL
Catalog No. 503073886
Encompass_Preferred
1mL dispense of HiCap T7 RNA Polymerase. Codex HiCap RNA Polymerase is a co-transcriptional capping RNA polymerase engineered to produce synthetic RNA to meet increasing demands for safety and cost-effectiveness. The HiCap enzyme generates fewer undesirable double-stranded RNA (dsRNA) byproducts than wild-type T7 (WT T7) RNA polymerase, reducing unwanted immunogenicity, while efficiently incorporating commercially available cap analogs.
Catalog No. 50-307-3886 Supplier Aldevron Supplier No. 91451ML
1mL dispense of HiCap T7 RNA Polymerase. Codex HiCap RNA Polymerase is a co-transcriptional capping RNA polymerase engineered to produce synthetic RNA to meet increasing demands for safety and cost-effectiveness. The HiCap enzyme generates fewer undesirable double-stranded RNA (dsRNA) byproducts than wild-type T7 (WT T7) RNA polymerase, reducing unwanted immunogenicity, while efficiently incorporating commercially available cap analogs.