Learn More
Medchemexpress LLC Codlemone (codlelure) | 33956-49-9 | 95.2% | 182.30 g/mol | C12H22O | 250MG
SDP
Catalog No. 5000236854
Encompass_Preferred
Codlemone (codlelure) is the primary sex pheromone of the codling moth and is supplied as a purified natural-product analytical standard and lure for entomology research, monitoring, and integrated pest management studies. It is appropriate for analytical reference, behavioral assays, and field trapping applications.
  • Primary sex pheromone for codling moth monitoring.
  • Suitable as an analytical standard for GC/MS and behavioral assays.
  • Approx. 95.2% purity.
  • Molecular formula C12H22O; molecular weight 182.30 g/mol.
  • Packaged as 250 mg and intended for refrigerated long-term storage.
Catalog No. 50-002-36854 Supplier Medchemexpress LLC Supplier No. HY124466250MG
Codlemone (codlelure) is the primary sex pheromone of the codling moth and is supplied as a purified natural-product analytical standard and lure for entomology research, monitoring, and integrated pest management studies. It is appropriate for analytical reference, behavioral assays, and field trapping applications.
  • Primary sex pheromone for codling moth monitoring.
  • Suitable as an analytical standard for GC/MS and behavioral assays.
  • Approx. 95.2% purity.
  • Molecular formula C12H22O; molecular weight 182.30 g/mol.
  • Packaged as 250 mg and intended for refrigerated long-term storage.