Learn More
Hach Company Chemical Oxygen Demand (COD) TNTplus Vial Test, LR (3-150 mg/L COD), 25 Tests
Supplier:  Hach Company TNT821
Encompass
Hach TNTplus COD vial test reagents are incredibly simple to use and cut down significantly on waste generation compared to other COD test kits. TNT821 is based on Hach Method 8000 for virtually any type of water sample. Method 8000 is USEPA approved for wastewater analysis under 40 CFR 136.3 and based on Standard Methods 5220 D. Hach TNTplus COD vial tests are available in multiple ranges. TNT821 provides a result in the Low Range of 3-150 mg/L COD and is often used for Chemical Oxygen Demand measurement in wastewater effluent. Hach TNT821 does not require a separate blanking vial for further waste reduction. Make the switch to TNTplus COD reagents today!For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion Method. Simply pipette in sample, digest, and read in your Hach Spectrophotometer. 25 tests per box.
Catalog No. 50-199-8113