Learn More
GA International Cody-3™ Printer Thermal Transfer/Direct Thermal
Catalog No. 5000819163
Encompass
GA International’s Cody-3 is a reliable thermal desktop printer that can print in thermal-transfer and direct thermal modes. Ideal for small to medium volume printing, the Cody-3 can print high-quality 300 dpi text with a 102 mm/s print speed, perfect for printing text, graphics, 1D- and 2D-barcodes. It provides a variety of connection options, including USB and Ethernet, for added flexibility and fast device connection. Bundled with GoLabel II software for unlimited, intuitive label and barcode design.
Catalog No. 50-008-19163 Supplier GA International Supplier No. CODY3
GA International’s Cody-3 is a reliable thermal desktop printer that can print in thermal-transfer and direct thermal modes. Ideal for small to medium volume printing, the Cody-3 can print high-quality 300 dpi text with a 102 mm/s print speed, perfect for printing text, graphics, 1D- and 2D-barcodes. It provides a variety of connection options, including USB and Ethernet, for added flexibility and fast device connection. Bundled with GoLabel II software for unlimited, intuitive label and barcode design.