Learn More
Medchemexpress LLC Coelenteramine 400a | 70217-82-2 | MFCD04114093 | 99.7% | 391.46 | C26H21N3O | 10 MG
SDP
Catalog No. 5000307744
Encompass_Preferred
Coelenteramine 400a is a coelenterazine-derived luciferase substrate used for Renilla luciferase assays, bioluminescence detection, and BRET studies. Supplied as a solid for research use, it is high-purity and should be protected from light during storage and handling.
  • Produces 395-400 nm emission with Renilla luciferase
  • High purity suitable for sensitive assays
  • Solid form that dissolves in common organic solvents
  • Recommended storage: -20°C, protect from light; in solvent: -80°C for long-term
  • Offered in research-scale quantities for assay development
Catalog No. 50-003-07744 Supplier Medchemexpress LLC Supplier No. HY11846210MG
Coelenteramine 400a is a coelenterazine-derived luciferase substrate used for Renilla luciferase assays, bioluminescence detection, and BRET studies. Supplied as a solid for research use, it is high-purity and should be protected from light during storage and handling.
  • Produces 395-400 nm emission with Renilla luciferase
  • High purity suitable for sensitive assays
  • Solid form that dissolves in common organic solvents
  • Recommended storage: -20°C, protect from light; in solvent: -80°C for long-term
  • Offered in research-scale quantities for assay development