Learn More
Medchemexpress LLC Coelenteramine 400a | 70217-82-2 | MFCD04114093 | 99.7% | 391.46 | C26H21N3O | 5 MG
SDP
Catalog No. 5000227439
Encompass_Preferred
Coelenteramine 400a is a coelenterazine derivative used as a Renilla luciferase substrate that shifts bioluminescence emission to approximately 395 nm, improving signal resolution in BRET and other RLuc-based assays.
  • Renilla luciferase substrate producing blue emission near 395 nm.
  • Enhances signal resolution for bioluminescence resonance energy transfer (BRET) assays.
  • High purity (99.7%) for reproducible results.
  • Available in small, research-scale quantities suitable for assay development.
  • Molecular formula C26H21N3O and molecular weight ~391.5 g/mol.
Catalog No. 50-002-27439 Supplier Medchemexpress LLC Supplier No. HY1184625MG
Coelenteramine 400a is a coelenterazine derivative used as a Renilla luciferase substrate that shifts bioluminescence emission to approximately 395 nm, improving signal resolution in BRET and other RLuc-based assays.
  • Renilla luciferase substrate producing blue emission near 395 nm.
  • Enhances signal resolution for bioluminescence resonance energy transfer (BRET) assays.
  • High purity (99.7%) for reproducible results.
  • Available in small, research-scale quantities suitable for assay development.
  • Molecular formula C26H21N3O and molecular weight ~391.5 g/mol.