Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coelenteramine 400a | 70217-82-2 | MFCD04114093 | 99.7% | 391.5 g/mol | C26H21N3O | 25 MG
SDP

Supplier:  Medchemexpress LLC HY11846225MG

Encompass_Preferred

Coelenteramine 400a is a coelenterazine-derived luciferase substrate used for Renilla luciferase (RLuc) assays and bioluminescence resonance energy transfer (BRET) research. It produces blue bioluminescent emission with a peak near 395 nm and is supplied as a solid with high reported purity.

  • Provides blue emission with peak near 395 nm.
  • Suitable for Renilla luciferase (RLuc) assays and BRET research.
  • High reported purity (99.7%).
  • Supplied as a solid; store protected from light at -20°C.
  • Available in small laboratory quantities (25 MG).
  • Follow safety data sheet precautions; harmful if swallowed and may irritate skin, eyes, and respiratory system.

Catalog No. 50-002-27618


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.