Learn More
Cayman Chemical Coelenterazine 400a, Purity: ≥95%, CAS: 70217-82-2, 5 mg
SDP
Catalog No. 502971824
Encompass_Preferred
A bisdeoxy derivative of coelenterazine that produces an emission maximum of 395 nm following conversion by Renilla luciferase; commonly used in BRET2 protocols to evaluate protein-protein interactions
Catalog No. 50-297-1824 Supplier Cayman Chemical Supplier No. 161575 MG
A bisdeoxy derivative of coelenterazine that produces an emission maximum of 395 nm following conversion by Renilla luciferase; commonly used in BRET2 protocols to evaluate protein-protein interactions