Learn More
Cayman Chemical CoelenterazIn h 500ug
SDP
Catalog No. 502966057
Encompass_Preferred
A synthetic derivative of native coelenterazine that exhibits 16-fold higher luminescence intensity (emission maximum ~466 nm; half-total time of 0.6-1.2 sec) than native coelenterazine
Catalog No. 50-296-6057 Supplier Cayman Chemical Supplier No. 16894500 UG
A synthetic derivative of native coelenterazine that exhibits 16-fold higher luminescence intensity (emission maximum ~466 nm; half-total time of 0.6-1.2 sec) than native coelenterazine