Learn More
Selleck Chemical LLC Coelenterazine 1mg 55779-48-1
SDP
Catalog No. 501365238
Encompass_Preferred
Coelenterazine is a luminescent enzyme substrate, used for monitoring reporter genes in BRET, ELISA and HTS techniques. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5238 Supplier Selleck Chemical LLC Supplier No. S77771MG
Coelenterazine is a luminescent enzyme substrate, used for monitoring reporter genes in BRET, ELISA and HTS techniques. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.