Learn More
Gold Biotechnology Inc Coelenterazine 400 a 1 mg
SDP
Catalog No. NC2542806
Encompass
Coelenterazine 400a is a derivative of coelenterazine and is a good substrate for Renilla reniformis luciferase (RLuc), but does not oxidize well with Gaussia (GLuc). It is the preferred substrate for BRET (Bioluminescence Resonance Energy Transfer) since its emission maximum of 400nm has minimal interference with the GFP emission.
Catalog No. NC2542806 Supplier Gold Biotechnology Inc Supplier No. C3201
Coelenterazine 400a is a derivative of coelenterazine and is a good substrate for Renilla reniformis luciferase (RLuc), but does not oxidize well with Gaussia (GLuc). It is the preferred substrate for BRET (Bioluminescence Resonance Energy Transfer) since its emission maximum of 400nm has minimal interference with the GFP emission.