Learn More
Medchemexpress LLC Coelenterazine 400a | 70217-82-2 | MFCD04114093 | 99.7% | 391.46 g/mol | C26H21N3O | 1 MG
SDP
Catalog No. 5000227617
Encompass_Preferred
Coelenterazine 400a is a bisdeoxy derivative of coelenterazine that serves as a blue-emitting substrate for Renilla luciferase (RLuc). It is used in bioluminescence and BRET/BRET2 assays to provide sensitive reporter signals in the ~395-400 nm range.
  • Blue emission with an approximate 395 nm peak, suitable for BRET/BRET2 studies.
  • High reported purity for reproducible assay results.
  • Appropriate for use as an RLuc substrate in luciferase reporter assays.
  • Available in small research quantities for assay development.
  • Synonymous with bisdeoxycoelenterazine and coelenteramine 400a.
Catalog No. 50-002-27617 Supplier Medchemexpress LLC Supplier No. HY1184621MG
Coelenterazine 400a is a bisdeoxy derivative of coelenterazine that serves as a blue-emitting substrate for Renilla luciferase (RLuc). It is used in bioluminescence and BRET/BRET2 assays to provide sensitive reporter signals in the ~395-400 nm range.
  • Blue emission with an approximate 395 nm peak, suitable for BRET/BRET2 studies.
  • High reported purity for reproducible assay results.
  • Appropriate for use as an RLuc substrate in luciferase reporter assays.
  • Available in small research quantities for assay development.
  • Synonymous with bisdeoxycoelenterazine and coelenteramine 400a.