Learn More
Medchemexpress LLC HY-18743 500ug , Coelenterazine CAS:55779-48-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18743500UG
Encompass_Preferred
HY-18743 500ug Coelenterazine CAS: 55779-48-1 Coelenterazine is a luminescent enzyme substrate for apoaequorin and Renilla luciferase. Renilla luciferase and substrate coelenterazine has been used as the bioluminescence donor in bioluminescence resonance energy transfer (BRET) to detect protein-protein interactions. Coelenterazine is a superoxide anion-sensitive chemiluminescent probe and it can also be used in chemiluminescent detection of peroxynitrite. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0223