Learn More
Medchemexpress LLC Coelenterazine | 55779-48-1 | 98.59% | 423.46 | 500 MG
SDP
Catalog No. 5000386065
Encompass_Preferred
Coelenterazine is a luminescent enzyme substrate for apoaequorin and Renilla luciferase. It has been used as the bioluminescence donor in bioluminescence resonance energy transfer (BRET) to detect protein-protein interactions. This compound is also a superoxide anion-sensitive chemiluminescent probe and can be used in chemiluminescent detection of peroxynitrite (Ex/Em = 429/466 nm).
  • Luminescent enzyme substrate for apoaequorin and Renilla luciferase.
  • Used as a bioluminescence donor in BRET to detect protein-protein interactions.
  • Superoxide anion-sensitive chemiluminescent probe.
  • Can be used in chemiluminescent detection of peroxynitrite.
Catalog No. 50-003-86065 Supplier Medchemexpress LLC Supplier No. HY18743500MG
Coelenterazine is a luminescent enzyme substrate for apoaequorin and Renilla luciferase. It has been used as the bioluminescence donor in bioluminescence resonance energy transfer (BRET) to detect protein-protein interactions. This compound is also a superoxide anion-sensitive chemiluminescent probe and can be used in chemiluminescent detection of peroxynitrite (Ex/Em = 429/466 nm).
  • Luminescent enzyme substrate for apoaequorin and Renilla luciferase.
  • Used as a bioluminescence donor in BRET to detect protein-protein interactions.
  • Superoxide anion-sensitive chemiluminescent probe.
  • Can be used in chemiluminescent detection of peroxynitrite.