Learn More
Selleck Chemical LLC Coelenterazine
SDP
Supplier:  Selleck Chemical LLC S7777100MG
Encompass_Preferred
Coelenterazine is a luminescent enzyme substrate for apoaequorin and Renilla luciferase Renilla luciferase and substrate coelenterazine have been used as bioluminescence donors in bioluminescence resonance energy transfer (BRET) to detect protein-protein interactions Coelenterazine is a superoxide anion-sensitive chemiluminescent probe and it can also be used in the chemiluminescent detection of peroxynitrite
Catalog No. 50-000-01003