Learn More
Alkali Scientific Coelenterazine H, 1 Mg
SDP
Catalog No. 5000660273
Encompass_Preferred
Coelenterazine H is a luminescent substrate widely used in biochemical assays, especially in studying the activity of luciferases. This 1mg reagent is used to trigger light emission in the presence of luciferase enzymes, making it valuable for applications in molecular biology, cell biology, and immunology. The light output provides a sensitive and quantifiable measure of enzymatic activity, making Coelenterazine H an essential tool for assay systems such as reporter gene assays and ATP measurements. Its ability to generate light makes it ideal for in vitro and in vivo imaging of biological processes, and it is commonly used in research related to cellular signaling, gene expression, and enzyme kinetics.
Catalog No. 50-006-60273 Supplier Alkali Scientific Supplier No. COE001CP
Coelenterazine H is a luminescent substrate widely used in biochemical assays, especially in studying the activity of luciferases. This 1mg reagent is used to trigger light emission in the presence of luciferase enzymes, making it valuable for applications in molecular biology, cell biology, and immunology. The light output provides a sensitive and quantifiable measure of enzymatic activity, making Coelenterazine H an essential tool for assay systems such as reporter gene assays and ATP measurements. Its ability to generate light makes it ideal for in vitro and in vivo imaging of biological processes, and it is commonly used in research related to cellular signaling, gene expression, and enzyme kinetics.