Learn More
Alkali Scientific Coelenterazine H, 10 Mg
SDP
Catalog No. 5000660281
Encompass_Preferred
Coelenterazine H, a luminescent substrate, is essential for bioluminescent assays involving luciferase enzymes. This 10mg reagent is used to catalyze light emission in the presence of luciferase, offering a sensitive method for measuring enzyme activity. The substrate is employed in a wide range of assays including gene expression studies, cellular signaling, and immunoassays. It provides real-time detection of biochemical processes, making it an important tool for researchers in molecular biology, cell biology, and pharmacology. Coelenterazine H is especially valuable in applications like reporter gene assays and ATP quantification, where accurate and reproducible light output is crucial for analysis.
Catalog No. 50-006-60281 Supplier Alkali Scientific Supplier No. COE002CP
Coelenterazine H, a luminescent substrate, is essential for bioluminescent assays involving luciferase enzymes. This 10mg reagent is used to catalyze light emission in the presence of luciferase, offering a sensitive method for measuring enzyme activity. The substrate is employed in a wide range of assays including gene expression studies, cellular signaling, and immunoassays. It provides real-time detection of biochemical processes, making it an important tool for researchers in molecular biology, cell biology, and pharmacology. Coelenterazine H is especially valuable in applications like reporter gene assays and ATP quantification, where accurate and reproducible light output is crucial for analysis.