Learn More
Eurofins Discoverx Coelenterazine h, 10 mg. (90-0084L)
Catalog No. 501999539
Encompass
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 10 mg of dry powder. For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.
Catalog No. 50-199-9539 Supplier Eurofins Discoverx Supplier No. 900084L
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 10 mg of dry powder. For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.