Learn More
Research Products International Corp Coelenterazine-H, 10 Milligrams
SDP
Supplier:  Research Products International Corp C615000.01
Encompass_Preferred
Highly sensitive and specific luniophore used for measuring changes in levels of superoxide and Ca2+ in cells transfected with apoaequorin cDNA. Exhibits 16-fold greater luminescence intensity as compared to native Coelenterazine.
Catalog No. 50-995-841