Learn More
TARGETMOL CHEMICALS INC Coelenterazine h 1MG
SDP
Catalog No. 502876407
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Coelenterazine H is a synthetic derivative of coelenterazine (a light-emitting biomolecule).Coelenterazine h (CLZN-h) is more sensitive to Ca 2+ than is the native complex, thus providing a valuable tool for measuring small changes in Ca 2+ concentrations. Purity 99.49%
Catalog No. 50-287-6407 Supplier TARGETMOL CHEMICALS INC Supplier No. T149941MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Coelenterazine H is a synthetic derivative of coelenterazine (a light-emitting biomolecule).Coelenterazine h (CLZN-h) is more sensitive to Ca 2+ than is the native complex, thus providing a valuable tool for measuring small changes in Ca 2+ concentrations. Purity 99.49%