Learn More
Eurofins Discoverx Coelenterazine h, 1 mg. (90-0084)
Catalog No. 501999538
Encompass
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 1 mg of dry powder. For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.
Catalog No. 50-199-9538 Supplier Eurofins Discoverx Supplier No. 900084
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 1 mg of dry powder. For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.