Learn More
Medchemexpress LLC Coelenterazine h (2-deoxycoelenterazine) | 50909-86-9 | 99.4% | 407.46 | C26H21N3O2 | 10 MG
SDP
Catalog No. 5000297559
Encompass_Preferred
Coelenterazine h (2-deoxycoelenterazine) is a yellow to orange solid luminescent substrate used in biochemical and cell-based assays. It functions as a substrate for Renilla luciferase, offers increased sensitivity to Ca2+ for detecting small calcium changes, and is supplied with high purity; the compound is unstable in solution and should be prepared fresh and stored protected from light at -20°C.
  • Is a luminescent substrate for Renilla luciferase, enabling bioluminescent assays.
  • Shows increased sensitivity to Ca2+, aiding detection of small calcium changes.
  • Supplied as a yellow to orange solid with high purity (≈99.4%).
  • Exhibits excitation/emission at 437/466 nm for assay matching.
  • Is unstable in solution; prepare fresh solutions and store at -20°C protected from light.
Catalog No. 50-002-97559 Supplier Medchemexpress LLC Supplier No. HYD102410MG
Coelenterazine h (2-deoxycoelenterazine) is a yellow to orange solid luminescent substrate used in biochemical and cell-based assays. It functions as a substrate for Renilla luciferase, offers increased sensitivity to Ca2+ for detecting small calcium changes, and is supplied with high purity; the compound is unstable in solution and should be prepared fresh and stored protected from light at -20°C.
  • Is a luminescent substrate for Renilla luciferase, enabling bioluminescent assays.
  • Shows increased sensitivity to Ca2+, aiding detection of small calcium changes.
  • Supplied as a yellow to orange solid with high purity (≈99.4%).
  • Exhibits excitation/emission at 437/466 nm for assay matching.
  • Is unstable in solution; prepare fresh solutions and store at -20°C protected from light.