Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coelenterazine h | 50909-86-9 | 98.8% | 10 MG
SDP

Catalog No. 5000297675
Encompass_Preferred

Coelenterazine h hydrochloride is a coelenterazine derivative used as a luminescent substrate for Renilla luciferase and related bioluminescent assays. It is suitable for sensitive luciferase detection and applications measuring Ca2+-dependent luminescence.

  • Luminescent substrate for Renilla luciferase and related enzymes.
  • High chemical purity (approximately 98.8%).
  • Molecular weight 443.92 and molecular formula C26H22ClN3O2.
  • Soluble in ethanol at ≥ 50 mg/mL.
  • Store protected from light, under nitrogen, at -20°C; stock solutions stable at -80°C for up to 6 months.
  • Supplied as a 10 MG vial for laboratory use.

Catalog No. 50-002-97675 Supplier Medchemexpress LLC Supplier No. HYD1024A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coelenterazine h hydrochloride is a coelenterazine derivative used as a luminescent substrate for Renilla luciferase and related bioluminescent assays. It is suitable for sensitive luciferase detection and applications measuring Ca2+-dependent luminescence.

  • Luminescent substrate for Renilla luciferase and related enzymes.
  • High chemical purity (approximately 98.8%).
  • Molecular weight 443.92 and molecular formula C26H22ClN3O2.
  • Soluble in ethanol at ≥ 50 mg/mL.
  • Store protected from light, under nitrogen, at -20°C; stock solutions stable at -80°C for up to 6 months.
  • Supplied as a 10 MG vial for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.