Learn More
Research Products International Corp Coelenterazine, Native, 10 Milligrams
SDP
Supplier:  Research Products International Corp C709000.01
Encompass_Preferred
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion. Used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA.
Catalog No. 50-995-845