Learn More
Regis Technologies Inc Coelenterazine, Native | 55779-48-1 | 10mg
SDP
Supplier:  Regis Technologies Inc 1361204200
Encompass
Coelenterazine. Regis Technologies manufactures Coelenterazine for use in research applications including reporter gene assays, ELISA assays, detection of calcium ions, whole animal assays and other biochemical assays. We produce high purity Coelenterazine to ensure the highest signal, lowest background, highest reproducibility, and lowest sensitivity levels for your application. Our manufacturing capabilities enable us to supply you with the highest purity material for your short term and long term needs, and the flexibility to help you overcome any obstacles. Inquire about our custom sizes and packaging.
Catalog No. NC0971570