Learn More
Research Products International Corp Coenzyme A, Free Acid, 25 Milligrams
SDP
Supplier:  Research Products International Corp C708000.025
Encompass_Preferred
Extracted from yeast. Coenzyme A is a coenzyme, notable for its role in the synthesis and oxidation of fatty acids, and the oxidation of pyuvate in the citric acid cycle.
Catalog No. 50-488-625