Learn More
Medchemexpress LLC Coenzyme A S-[2-[[(5S)-5-(acetylamino)-6-amino-6-oxohexyl]amino]-2-oxoethyl] trifluoroacetic acid | 00-00-0 | 98.5% | C33H54F3N10O21P3S | 10MG
SDP
Catalog No. 5000261718
Encompass_Preferred
Lys-CoA TFA (Lys-coenzyme A) is a selective inhibitor of the p300 histone acetyltransferase used as a biochemical tool to study p300-mediated acetylation and transcriptional regulation. It exhibits nanomolar potency against p300 and marked selectivity over related acetyltransferases, and is supplied as a high-purity solid for laboratory research.
  • Selective p300 HAT inhibitor with p300 IC50 50-500 nM
  • Greater than 100-fold selectivity over PCAF
  • High purity (98.5%) for research applications
  • Solid, white to off-white physical form
  • Recommended sealed storage at -20°C; in solvent store at -80°C for long-term
Catalog No. 50-002-61718 Supplier Medchemexpress LLC Supplier No. HY13103510MG
Lys-CoA TFA (Lys-coenzyme A) is a selective inhibitor of the p300 histone acetyltransferase used as a biochemical tool to study p300-mediated acetylation and transcriptional regulation. It exhibits nanomolar potency against p300 and marked selectivity over related acetyltransferases, and is supplied as a high-purity solid for laboratory research.
  • Selective p300 HAT inhibitor with p300 IC50 50-500 nM
  • Greater than 100-fold selectivity over PCAF
  • High purity (98.5%) for research applications
  • Solid, white to off-white physical form
  • Recommended sealed storage at -20°C; in solvent store at -80°C for long-term