Learn More
Cayman Chemical Coenzyme A sodium salt hydra
SDP
Supplier:  Cayman Chemical 2172225 MG
Encompass_Preferred
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the coenzyme A biosynthetic pathway has been identified as a target for the development of antibacterial compounds
Catalog No. 50-298-5972