Learn More
Cayman Chemical Coenzyme A (sodium salt), Purity: ≥90%, CAS: 55672-92-9, 50 mg
SDP
Catalog No. 502987345
Encompass_Preferred
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the coenzyme A biosynthetic pathway has been identified as a target for the development of antibacterial compounds
Catalog No. 50-298-7345 Supplier Cayman Chemical Supplier No. 2172250 MG
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the coenzyme A biosynthetic pathway has been identified as a target for the development of antibacterial compounds