Learn More
Sigma Aldrich Fine Chemicals Biosciences Coenzyme A sodum salt hyd100MG
Catalog No. 502903572
Encompass
Coenzyme A (CoA) is an essential cofactor in living systems and is synthesized from pantothenic acid (vitamin B5) The CoA levels in mitochondria and peroxisomes correspond to 2-5 mM and 0.7 mM respectively. Cytosolic CoA is in the range of 0.05 mM to 0.14 mM
Catalog No. 50-290-3572 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C3144100MG
Coenzyme A (CoA) is an essential cofactor in living systems and is synthesized from pantothenic acid (vitamin B5) The CoA levels in mitochondria and peroxisomes correspond to 2-5 mM and 0.7 mM respectively. Cytosolic CoA is in the range of 0.05 mM to 0.14 mM