Learn More
Research Products International Corp Coenzyme A Trilithium Salt 1 G
SDP
Catalog No. 502230308
Encompass_Preferred
Coenzyme A (CoA, CoASH, HSCoA) is a coenzyme that facilitates enzymatic acyl-group transfer reactions and supports the synthesis and oxidation of fatty acids. CoA is involved in the mechanisms of a wide variety of enzymes.
Catalog No. 50-223-0308 Supplier Research Products International Corp Supplier No. C952751.0
Coenzyme A (CoA, CoASH, HSCoA) is a coenzyme that facilitates enzymatic acyl-group transfer reactions and supports the synthesis and oxidation of fatty acids. CoA is involved in the mechanisms of a wide variety of enzymes.