Learn More
Research Products International Corp Coenzyme A Trilithium Salt [Acetyl Coenzyme A, Trilithium Salt], 100 Milligrams
SDP
Catalog No. 501368212
Encompass_Preferred
Coenzyme A (CoA, CoASH, HSCoA) is a coenzyme that facilitates enzymatic acyl-group transfer reactions and supports the synthesis and oxidation of fatty acids. CoA is involved in the mechanisms of a wide variety of enzymes.
Catalog No. 50-136-8212 Supplier Research Products International Corp Supplier No. C952750.1
Coenzyme A (CoA, CoASH, HSCoA) is a coenzyme that facilitates enzymatic acyl-group transfer reactions and supports the synthesis and oxidation of fatty acids. CoA is involved in the mechanisms of a wide variety of enzymes.