Learn More
Research Products International Corp Coenzyme A Trilithium Salt, 25 MG
SDP
Catalog No. 501977943
Encompass_Preferred
Coenzyme A (CoA, CoASH, HSCoA) is a coenzyme that facilitates enzymatic acyl-group transfer reactions and supports the synthesis and oxidation of fatty acids. CoA is involved in the mechanisms of a wide variety of enzymes.
Catalog No. 50-197-7943 Supplier Research Products International Corp Supplier No. C952750.025
Coenzyme A (CoA, CoASH, HSCoA) is a coenzyme that facilitates enzymatic acyl-group transfer reactions and supports the synthesis and oxidation of fatty acids. CoA is involved in the mechanisms of a wide variety of enzymes.