Learn More
Sigma Aldrich Fine Chemicals Biosciences Coenzyme A Trilithium Salt | ≥85% Purity | Mol Wt: 767.5 | ®93-18-4-3 | 10 mg
Catalog No. NC2669001
Encompass
Coenzyme A trilithium salt | Purity: ≥85% | Mol Wt: 767.5 | CAS No: 93-18-4-3 | Biochemical cofactor involved in metabolic pathways and energy production via acyl transfer and thioesterification | 10mg
Catalog No. NC2669001 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C301910MG
Coenzyme A trilithium salt | Purity: ≥85% | Mol Wt: 767.5 | CAS No: 93-18-4-3 | Biochemical cofactor involved in metabolic pathways and energy production via acyl transfer and thioesterification | 10mg