Learn More
Cayman Chemical Coenzyme A (lithium salt hydrate), Purity: ≥90%, 250 mg
SDP
Catalog No. 502964014
Encompass_Preferred
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the CoA biosynthetic pathway has been identified as a target for the development of antibacterial compounds
Catalog No. 50-296-4014 Supplier Cayman Chemical Supplier No. 21499250 MG
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the CoA biosynthetic pathway has been identified as a target for the development of antibacterial compounds