Learn More
Cayman Chemical Coenzyme AsodIM salt hydra 5mg
SDP
Catalog No. 502987372
Encompass_Preferred
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the coenzyme A biosynthetic pathway has been identified as a target for the development of antibacterial compounds
Catalog No. 50-298-7372 Supplier Cayman Chemical Supplier No. 217225 MG
An essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism; the pantothenate kinase step of the coenzyme A biosynthetic pathway has been identified as a target for the development of antibacterial compounds