Learn More
TARGETMOL CHEMICALS INC Coenzyme FO 1MG
SDP
Catalog No. 502879080
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Coenzyme FO, a deazaflavin chromophore, is essential as a hydride acceptor/donor in the central methanogenic pathway [1][2]. Purity 97.66%
Catalog No. 50-287-9080 Supplier TARGETMOL CHEMICALS INC Supplier No. T379781MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Coenzyme FO, a deazaflavin chromophore, is essential as a hydride acceptor/donor in the central methanogenic pathway [1][2]. Purity 97.66%