Learn More
Medchemexpress LLC HY-136497 5mg Medchemexpress, Coenzyme FO CAS:37333-48-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364975MG
Encompass_Preferred
Medchemexpress, HY-136497 5mg Coenzyme FO CAS:37333-48-5 Coenzyme FO, a deazaflavin chromophore, acts as an important hydride acceptor/donor in the central methanogenic pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1445